Anti-KLK10 Mouse Monoclonal Antibody [clone: 1G8]

Lieferant: United States Biological
USBI128709EA 733 CHF
USBI128709
Anti-KLK10 Mouse Monoclonal Antibody [clone: 1G8]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: KLK10 (kallikrein-related peptidase 10)
Clonality: monoclonal
Clone: 1G8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Lambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR