Anti-KLK10 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI128708EA 797 CHF
USBI128708
Anti-KLK10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLK10 (kallikrein-related peptidase 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR