Anti-KAL1 Mouse Monoclonal Antibody [clone: 2E8]

Supplier: US Biological
USBI128706EA 702 CHF
USBI128706
Anti-KAL1 Mouse Monoclonal Antibody [clone: 2E8]
Antibodies
Type: Primary
Antigen: KAL1 (Kallmann syndrome 1 sequence)
Clonality: monoclonal
Clone: 200000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR