Anti-AIMP2 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI128690EA 733 CHF
USBI128690
Anti-AIMP2 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: AIMP2 (aminoacyl tRNA synthetase complex-interacting multifunctional protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR