Anti-JRKL Mouse Monoclonal Antibody [clone: 4E10]

Supplier: US Biological
USBI128688EA 702 CHF
USBI128688
Anti-JRKL Mouse Monoclonal Antibody [clone: 4E10]
Antibodies
Type: Primary
Antigen: JRKL (jerky homolog-like (mouse))
Clonality: monoclonal
Clone: 40000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR