Anti-JPH1 Mouse Monoclonal Antibody [clone: 2E6]

Supplier: US Biological
USBI128686EA 702 CHF
USBI128686
Anti-JPH1 Mouse Monoclonal Antibody [clone: 2E6]
Antibodies
Type: Primary
Antigen: JPH1 (junctophilin 1)
Clonality: monoclonal
Clone: 2000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR