Anti-JMY Mouse Monoclonal Antibody [clone: 2D7]

Supplier: US Biological
USBI128685EA 702 CHF
USBI128685
Anti-JMY Mouse Monoclonal Antibody [clone: 2D7]
Antibodies
Type: Primary
Antigen: JMY (junction mediating and regulatory protein, p53 cofactor)
Clonality: monoclonal
Clone: 2D7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR