Anti-KDM8 Mouse Monoclonal Antibody [clone: 1F3]

Supplier: US Biological
USBI128684EA 702 CHF
USBI128684
Anti-KDM8 Mouse Monoclonal Antibody [clone: 1F3]
Antibodies
Type: Primary
Antigen: KDM8 (lysine (K)-specific demethylase 8)
Clonality: monoclonal
Clone: 1F3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR