Anti-JMJD1C Mouse Monoclonal Antibody [clone: 5F12]

Supplier: US Biological
USBI128682EA 702 CHF
USBI128682
Anti-JMJD1C Mouse Monoclonal Antibody [clone: 5F12]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: JMJD1C (jumonji domain containing 1C)
Clonality: monoclonal
Clone: 5F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR