Anti-JAZF1 Mouse Polyclonal Antibody

Supplier: US Biological
USBI128680EA 702 CHF
USBI128680
Anti-JAZF1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: JAZF1 (JAZF zinc finger 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR