Anti-KDM5B Mouse Monoclonal Antibody [clone: 1G10]

Supplier: US Biological
USBI128679EA 702 CHF
USBI128679
Anti-KDM5B Mouse Monoclonal Antibody [clone: 1G10]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 1 ?g/ml

Type: Primary
Antigen: KDM5B (lysine (K)-specific demethylase 5B)
Clonality: monoclonal
Clone: 1G10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR