Anti-KDM5A Mouse Monoclonal Antibody [clone: 1H2]

Supplier: US Biological
USBI128678EA 702 CHF
USBI128678
Anti-KDM5A Mouse Monoclonal Antibody [clone: 1H2]
Antibodies
Type: Primary
Antigen: KDM5A (lysine (K)-specific demethylase 5A)
Clonality: monoclonal
Clone: 1H2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR