Anti-JAM2 Mouse Monoclonal Antibody [clone: 1G4]

Supplier: US Biological
USBI128677EA 702 CHF
USBI128677
Anti-JAM2 Mouse Monoclonal Antibody [clone: 1G4]
Antibodies
Recommended dilution: Immunochemistry: 1 ?g/ml

Type: Primary
Antigen: JAM2 (junctional adhesion molecule 2)
Clonality: monoclonal
Clone: 1G4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR