Anti-JAK3 Mouse Monoclonal Antibody [clone: 1E10]

Supplier: US Biological
USBI128675EA 702 CHF
USBI128675
Anti-JAK3 Mouse Monoclonal Antibody [clone: 1E10]
Antibodies
Type: Primary
Antigen: JAK3 (Janus kinase 3)
Clonality: monoclonal
Clone: 10000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR