Anti-JAG2 Mouse Monoclonal Antibody [clone: 4F10]

Supplier: US Biological
USBI128674EA 702 CHF
USBI128674
Anti-JAG2 Mouse Monoclonal Antibody [clone: 4F10]
Antibodies
Type: Primary
Antigen: JAG2 (jagged 2)
Clonality: monoclonal
Clone: 4F10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR