Anti-JAG1 Mouse Monoclonal Antibody [clone: 1E12]

Supplier: US Biological
USBI128673EA 702 CHF
USBI128673
Anti-JAG1 Mouse Monoclonal Antibody [clone: 1E12]
Antibodies
Type: Primary
Antigen: JAG1 (jagged 1)
Clonality: monoclonal
Clone: 1000000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR