Anti-ITPR1 Mouse Monoclonal Antibody [clone: 2B6]

Supplier: US Biological
USBI128668EA 702 CHF
USBI128668
Anti-ITPR1 Mouse Monoclonal Antibody [clone: 2B6]
Antibodies
Type: Primary
Antigen: ITPR1 (inositol 1,4,5-trisphosphate receptor, type 1)
Clonality: monoclonal
Clone: 2B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR