Anti-ITPKB Mouse Monoclonal Antibody [clone: 2F8]

Supplier: US Biological
USBI128666EA 702 CHF
USBI128666
Anti-ITPKB Mouse Monoclonal Antibody [clone: 2F8]
Antibodies
Type: Primary
Antigen: ITPKB (inositol-trisphosphate 3-kinase B)
Clonality: monoclonal
Clone: 2F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR