Anti-ITPK1 Mouse Monoclonal Antibody [clone: 2C8]

Supplier: US Biological
USBI128664EA 702 CHF
USBI128664
Anti-ITPK1 Mouse Monoclonal Antibody [clone: 2C8]
Antibodies
Type: Primary
Antigen: ITPK1 (inositol-tetrakisphosphate 1-kinase)
Clonality: monoclonal
Clone: 2C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR