Anti-ITPA Mouse Monoclonal Antibody [clone: 2H8]

Supplier: US Biological
USBI128660EA 702 CHF
USBI128660
Anti-ITPA Mouse Monoclonal Antibody [clone: 2H8]
Antibodies
Type: Primary
Antigen: ITPA (inosine triphosphatase (nucleoside triphosphate pyrophosphatase))
Clonality: monoclonal
Clone: 2H8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR