Drucken

Anti-ITM2B Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI128657EA 750 CHF
USBI128657
Anti-ITM2B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ITM2B (integral membrane protein 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR