Anti-STT3A Mouse Monoclonal Antibody [clone: 4D4]

Supplier: US Biological
USBI128655EA 702 CHF
USBI128655
Anti-STT3A Mouse Monoclonal Antibody [clone: 4D4]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: STT3A (STT3A, subunit of the oligosaccharyltransferase complex (catalytic))
Clonality: monoclonal
Clone: 4D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR