Anti-ITK Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128654EA 750 CHF
USBI128654
Anti-ITK Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ITK (IL2-inducible T-cell kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR