Anti-ITGB7 Mouse Monoclonal Antibody [clone: 8D3]

Supplier: US Biological
USBI128647EA 702 CHF
USBI128647
Anti-ITGB7 Mouse Monoclonal Antibody [clone: 8D3]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: ITGB7 (integrin, beta 7)
Clonality: monoclonal
Clone: 8D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR