Anti-ITGB7 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128646EA 750 CHF
USBI128646
Anti-ITGB7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ITGB7 (integrin, beta 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR