Anti-ITGB6 Mouse Monoclonal Antibody [clone: 4C3]

Supplier: US Biological
USBI128644EA 702 CHF
USBI128644
Anti-ITGB6 Mouse Monoclonal Antibody [clone: 4C3]
Antibodies
Type: Primary
Antigen: ITGB6 (integrin, beta 6)
Clonality: monoclonal
Clone: 4C3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR