Anti-ITGB5 Mouse Monoclonal Antibody [clone: 2C4]

Supplier: US Biological
USBI128641EA 702 CHF
USBI128641
Anti-ITGB5 Mouse Monoclonal Antibody [clone: 2C4]
Antibodies
Type: Primary
Antigen: ITGB5 (integrin, beta 5)
Clonality: monoclonal
Clone: 2C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR