Anti-ITGB3BP Mouse Monoclonal Antibody [clone: 3F6]

Supplier: US Biological
USBI128639EA 702 CHF
USBI128639
Anti-ITGB3BP Mouse Monoclonal Antibody [clone: 3F6]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: ITGB3BP (integrin beta 3 binding protein (beta3-endonexin))
Clonality: monoclonal
Clone: 3F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR