Anti-ITGB2 Mouse Monoclonal Antibody [clone: 5F12]

Supplier: US Biological
USBI128638EA 702 CHF
USBI128638
Anti-ITGB2 Mouse Monoclonal Antibody [clone: 5F12]
Antibodies
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: monoclonal
Clone: 5F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR