Anti-ITGB2 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128637EA 750 CHF
USBI128637
Anti-ITGB2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR