Anti-ITGB1BP2 Mouse Monoclonal Antibody [clone: 3G9]

Supplier: US Biological
USBI128634EA 702 CHF
USBI128634
Anti-ITGB1BP2 Mouse Monoclonal Antibody [clone: 3G9]
Antibodies
Type: Primary
Antigen: ITGB1BP2 (integrin beta 1 binding protein (melusin) 2)
Clonality: monoclonal
Clone: 3G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR