Anti-ITGAE (Integrin alpha E, CD103) Mouse Monoclonal Antibody [clone: 6B8]
Lieferant: United States Biological
USBI128631EA 733 CHF
USBI128631
Anti-ITGAE (Integrin alpha E, CD103) Mouse Monoclonal Antibody [clone: 6B8]
Antikörper
Type: Primary
Antigen: ITGAE (Integrin alpha E, CD103)
Clonality: Monoclonal
Clone: 6B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human