Anti-ITGA8 Mouse Monoclonal Antibody [clone: 2G7]

Supplier: US Biological
USBI128630EA 702 CHF
USBI128630
Anti-ITGA8 Mouse Monoclonal Antibody [clone: 2G7]
Antibodies
Type: Primary
Antigen: ITGA8 (integrin, alpha 8)
Clonality: monoclonal
Clone: 2G7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR