Anti-ITGA7 Mouse Monoclonal Antibody [clone: 8G2]

Supplier: US Biological
USBI128629EA 702 CHF
USBI128629
Anti-ITGA7 Mouse Monoclonal Antibody [clone: 8G2]
Antibodies
Type: Primary
Antigen: ITGA7 (integrin, alpha 7)
Clonality: monoclonal
Clone: 8G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR