Anti-ITGA4 Mouse Monoclonal Antibody [clone: 2C11]

Supplier: US Biological
USBI128625EA 702 CHF
USBI128625
Anti-ITGA4 Mouse Monoclonal Antibody [clone: 2C11]
Antibodies
Type: Primary
Antigen: ITGA4 (integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor))
Clonality: monoclonal
Clone: 2C11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR