Anti-ITGA4 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128624EA 750 CHF
USBI128624
Anti-ITGA4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ITGA4 (integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR