Anti-ITGA2 Mouse Monoclonal Antibody [clone: 2B6]

Supplier: US Biological
USBI128623EA 702 CHF
USBI128623
Anti-ITGA2 Mouse Monoclonal Antibody [clone: 2B6]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: ITGA2 (integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor))
Clonality: monoclonal
Clone: 2B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR