Anti-ITGA1 Mouse Monoclonal Antibody [clone: 4G11]

Lieferant: United States Biological
USBI128622EA 733 CHF
USBI128622
Anti-ITGA1 Mouse Monoclonal Antibody [clone: 4G11]
Antikörper
Type: Primary
Antigen: ITGA1 (integrin, alpha 1)
Clonality: monoclonal
Clone: 4G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR