Anti-ITCH Mouse Monoclonal Antibody [clone: 1B8]

Supplier: US Biological
USBI128616EA 702 CHF
USBI128616
Anti-ITCH Mouse Monoclonal Antibody [clone: 1B8]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: ITCH (itchy E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: 1B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR