Anti-ISL2 Mouse Monoclonal Antibody [clone: 1D9]

Supplier: US Biological
USBI128611EA 702 CHF
USBI128611
Anti-ISL2 Mouse Monoclonal Antibody [clone: 1D9]
Antibodies
Type: Primary
Antigen: ISL2 (ISL LIM homeobox 2)
Clonality: monoclonal
Clone: 1D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR