Anti-ISG20 Mouse Monoclonal Antibody [clone: 1B2-3C9]

Lieferant: United States Biological
USBI128605EA 733 CHF
USBI128605
Anti-ISG20 Mouse Monoclonal Antibody [clone: 1B2-3C9]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: ISG20 (interferon stimulated exonuclease gene 20kDa)
Clonality: monoclonal
Clone: 1B2-3C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR