Anti-ISG15 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI128601EA 788 CHF
USBI128601
Anti-ISG15 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ISG15 (ISG15 ubiquitin-like modifier)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR