Anti-IRX6 Mouse Monoclonal Antibody [clone: 1A5]

Supplier: US Biological
USBI128598EA 702 CHF
USBI128598
Anti-IRX6 Mouse Monoclonal Antibody [clone: 1A5]
Antibodies
Type: Primary
Antigen: IRX6 (iroquois homeobox 6)
Clonality: monoclonal
Clone: 1A5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR