Anti-IRX5 Mouse Monoclonal Antibody [clone: 6B5]

Supplier: US Biological
USBI128596EA 702 CHF
USBI128596
Anti-IRX5 Mouse Monoclonal Antibody [clone: 6B5]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: IRX5 (iroquois homeobox 5)
Clonality: monoclonal
Clone: 6B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR