Anti-IRX3 Mouse Monoclonal Antibody [clone: 3D8]

Supplier: US Biological
USBI128595EA 702 CHF
USBI128595
Anti-IRX3 Mouse Monoclonal Antibody [clone: 3D8]
Antibodies
Recommended dilution: Immunofluorescence: 30 ?g/ml

Type: Primary
Antigen: IRX3 (iroquois homeobox 3)
Clonality: monoclonal
Clone: 3D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR