Anti-IRS4 Mouse Monoclonal Antibody [clone: 5B9]

Supplier: US Biological
USBI128592EA 702 CHF
USBI128592
Anti-IRS4 Mouse Monoclonal Antibody [clone: 5B9]
Antibodies
Type: Primary
Antigen: IRS4 (insulin receptor substrate 4)
Clonality: monoclonal
Clone: 5B9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR