Anti-IRF3 Mouse Monoclonal Antibody [clone: 3C8]

Supplier: US Biological
USBI128587EA 702 CHF
USBI128587
Anti-IRF3 Mouse Monoclonal Antibody [clone: 3C8]
Antibodies
Type: Primary
Antigen: IRF3 (interferon regulatory factor 3)
Clonality: monoclonal
Clone: 3C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR