Anti-IRF2BP1 Mouse Polyclonal Antibody

Supplier: US Biological
USBI128583EA 702 CHF
USBI128583
Anti-IRF2BP1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IRF2BP1 (interferon regulatory factor 2 binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR