Anti-IRF2 Mouse Monoclonal Antibody [clone: 3B5]

Lieferant: United States Biological
USBI128582EA 733 CHF
USBI128582
Anti-IRF2 Mouse Monoclonal Antibody [clone: 3B5]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: IRF2 (interferon regulatory factor 2)
Clonality: monoclonal
Clone: 3B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR