Anti-IRF1 Mouse Monoclonal Antibody [clone: 2E4]

Supplier: US Biological
USBI128580EA 702 CHF
USBI128580
Anti-IRF1 Mouse Monoclonal Antibody [clone: 2E4]
Antibodies
Type: Primary
Antigen: IRF1 (interferon regulatory factor 1)
Clonality: monoclonal
Clone: 20000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR